3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-5(6)-carboxylic acid 98%

Supplier: Ambeed
Warning

A415439-1G A415439-25G A415439-5G
76773-836EA 28.65 USD
76773-836 76773-838 77626-832
3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-5(6)-carboxylic acid 98%
3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-5(6)-carboxylic acid

Precaución: for Research Use Only

Formula: C₄₂H₂₄O₁₄
MW: 752.63 g/mol
MDL Number: MFCD00151081
CAS Number: 1171493-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR