5-Chloro-8-nitroquinoline 95%

Supplier: Ambeed
Warning

A414623-100MG A414623-5G A414623-250MG
76773-830EA 22.28 USD
76773-830 77568-836 77626-830
5-Chloro-8-nitroquinoline 95%
5-Chloro-8-nitroquinoline

Precaución: For research use only.

Formula: C₉H₅ClN₂O₂
MW: 208.60 g/mol
MDL Number: MFCD00552691
CAS Number: 6942-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR