3-(Carbobenzoxyamino)-1-propanol 98%

Supplier: Ambeed
Warning

Synonyms: Benzyl N-(3-Hydroxypropyl)carbamate, N-(3-Hydroxypropyl)carbamic Acid Benzyl Ester, 3-(Cbz-amino)-1-propanol, Benzyl (3-hydroxypropyl)carbamate

A414218-1G A414218-25G A414218-5G A414218-100G A414218-500G
76773-824EA 6 USD
76773-824 76773-826 76773-828 77598-474 77878-163
3-(Carbobenzoxyamino)-1-propanol 98%
3-(Carbobenzoxyamino)-1-propanol

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO₃
MW: 209.25 g/mol
MDL Number: MFCD01321351
CAS Number: 34637-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR