Nickel(II) hexafluoroacetylacetonate hydrate 98%

Supplier: Ambeed
Danger

Synonyms: Hexafluoroacetylacetono Nickel(II) Salt, Bis(hexafluoroacetylacetonato)nickel(II) hydrate

A412278-250MG A412278-5G A412278-1G
76773-800EA 15.92 USD
76773-800 76773-802 77598-472
Nickel(II) hexafluoroacetylacetonate hydrate 98%
Nickel(II) hexafluoroacetylacetonate hydrate

Precaución: for Research Use Only

Formula: C₁₀H₂F₁₂NiO₄·xH₂O
MW: 472.80 g/mol (anhy)
MDL Number: MFCD00067506
CAS Number: 14949-69-0
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR