N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic acid 98%

Supplier: Ambeed
Warning

Synonyms: N-Fmoc-L-aspartic Acid, Fmoc-Asp-OH

A409401-100G A409401-25G A409401-500G
76773-788EA 75 USD
76773-788 77572-268 77878-158
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic acid 98%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic acid

Precaución: for Research Use Only

Formula: C₁₉H₁₇NO₆
MDL Number: MFCD00237654
CAS Number: 119062-05-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR