2-Chloro-6-(methylamino)pyrazine 98%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-N-methylpyrazin-2-amine

A407579-1G A407579-25G A407579-5G
76773-764EA 16.98 USD
76773-764 76773-766 76773-768
2-Chloro-6-(methylamino)pyrazine 98%
2-Chloro-6-(methylamino)pyrazine

Precaución: for Research Use Only

Formula: C₅H₆ClN₃
MW: 143.5700 g/mol
MDL Number: MFCD09878383
CAS Number: 848366-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR