5-Amino-2-(2,4-dichlorophenoxy)pyridine 97%

Supplier: Ambeed
Warning

A405136-1G A405136-250MG A405136-25G A405136-5G
76773-754EA 27 USD
76773-754 76773-756 76773-758 76773-760
5-Amino-2-(2,4-dichlorophenoxy)pyridine 97%
5-Amino-2-(2,4-dichlorophenoxy)pyridine

Precaución: for Research Use Only

Formula: C₁₁H₈Cl₂N₂O
MDL Number: MFCD00052643
CAS Number: 85686-48-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR