Methyl-5-hexenoate 98%

Supplier: Ambeed
Danger

Synonyms: Methyl 5-hexenoate, 5-Hexenoic Acid Methyl Ester

A404788-25G A404788-5G
76773-750EA 460.44 USD
76773-750 76773-752
Methyl-5-hexenoate 98%
Methyl-5-hexenoate

Precaución: for Research Use Only

Formula: C₇H₁₂O₂
MW: 128.17 g/mol
MDL Number: MFCD00671538
CAS Number: 2396-80-7
UN: 3272
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR