4-Bromo-3-fluorotoluene 98%

Supplier: Ambeed
Danger

A402226-10G A402226-25G A402226-5G A402226-100G
76773-736EA 10 USD
76773-736 76773-738 76773-740 77878-142
4-Bromo-3-fluorotoluene 98%
4-Bromo-3-fluorotoluene

Precaución: for Research Use Only

Formula: C₇H₆BrF
MDL Number: MFCD00040940
CAS Number: 452-74-4
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR