5-Bromo-3-ethyloxindole 97%

Supplier: Ambeed
Warning

A401226-100MG A401226-1G A401226-250MG
76773-724EA 51 USD
76773-724 76773-726 76773-728
5-Bromo-3-ethyloxindole 97%
5-Bromo-3-ethyloxindole

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrNO
MDL Number: MFCD06245570
CAS Number: 304876-05-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR