5-Ethyl-2-thiophenecarboxaldehyde 97%

Supplier: Ambeed
Warning

A401082-1G A401082-25G A401082-5G
76773-718EA 12 USD
76773-718 76773-720 76773-722
5-Ethyl-2-thiophenecarboxaldehyde 97%
5-Ethyl-2-thiophenecarboxaldehyde

Precaución: for Research Use Only

Formula: C₇H₈OS
MDL Number: MFCD00143457
CAS Number: 36880-33-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR