5,6-Diamino-2,3-dicyanopyrazine 98%

Supplier: Ambeed
Danger

Synonyms: 5,6-Diamino-2,3-pyrazinedicarbonitrile, 5,6-Diaminopyrazine-2,3-dicarbonitrile

A399314-100MG A399314-250MG A399314-1G
76773-698EA 37.14 USD
76773-698 76773-700 77626-814
5,6-Diamino-2,3-dicyanopyrazine 98%
5,6-Diamino-2,3-dicyanopyrazine

Precaución: for Research Use Only

Formula: C₆H₄N₆
MW: 160.14 g/mol
MDL Number: MFCD00191418
CAS Number: 36023-58-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR