2-Amino-4-(4-bromophenyl)-5-methylthiazole 98%

Supplier: Ambeed
Danger

Synonyms: 4-(4-Bromophenyl)-5-methylthiazol-2-amine

A396288-1G A396288-25G A396288-5G
76773-688EA 49.87 USD
76773-688 76773-690 76773-692
2-Amino-4-(4-bromophenyl)-5-methylthiazole 98%
2-Amino-4-(4-bromophenyl)-5-methylthiazole

Precaución: for Research Use Only

Formula: C₁₀H₉BrN₂S
MW: 269.16 g/mol
MDL Number: MFCD00170268
CAS Number: 65705-44-4
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR