5-Chloro-2-nitrobenzonitrile 98%

Supplier: Ambeed
Warning

A395676-1G A395676-25G A395676-5G
76773-682EA 13.8 USD
76773-682 76773-684 76773-686
5-Chloro-2-nitrobenzonitrile 98%
5-Chloro-2-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃ClN₂O₂
MW: 182.57 g/mol
MDL Number: MFCD00007286
CAS Number: 34662-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR