2-Methanesulfonyl-4,6-dimethoxypyrimidine 97%

Supplier: Ambeed
Warning

Synonyms: 4,6-Dimethoxy-2-(methylsulphonyl)pyrimidine

A395114-1000G A395114-100G A395114-10G A395114-500G
76773-674EA 108 USD
76773-674 76773-676 76773-678 76773-680
2-Methanesulfonyl-4,6-dimethoxypyrimidine 97%
2-Methanesulfonyl-4,6-dimethoxypyrimidine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₄S
MW: 218.23 g/mol
MDL Number: MFCD00672151
CAS Number: 113583-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR