5-Bromo-4,6-difluoropyrimidine 95%

Supplier: Ambeed
Danger

A394649-1G A394649-250MG
76773-670EA 120 USD
76773-670 76773-672
5-Bromo-4,6-difluoropyrimidine 95%
5-Bromo-4,6-difluoropyrimidine

Precaución: for Research Use Only

Formula: C₄HBrF₂N₂
MDL Number: MFCD08686978
CAS Number: 946681-88-5
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR