2-Amino-6-fluorophenol 97%

Supplier: Ambeed
Warning

Synonyms: 6-Fluoro-2-aminophenol

A394551-100MG A394551-1G A394551-5G
76773-666EA 8.49 USD
76773-666 76773-668 77626-804
2-Amino-6-fluorophenol 97%
2-Amino-6-fluorophenol

Precaución: for Research Use Only

Formula: C₆H₆FNO
MW: 127.12 g/mol
MDL Number: MFCD08532465
CAS Number: 53981-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR