Boc-Gly-ONp 98%

Supplier: Ambeed
Warning

A390709-100G A390709-25G
76773-646EA 266.29 USD
76773-646 76773-648
Boc-Gly-ONp 98%
Boc-Gly-ONp

Precaución: for Research Use Only

Formula: C₁₃H₁₆N₂O₆
MW: 296.279 g/mol
MDL Number: MFCD00038125
CAS Number: 3655-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR