5-Methyl-1-phenyl-1H-pyrazole-4-boronic acid, pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 5-Methyl-1-phenyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 2-(5-Methyl-1-phenyl-1H-pyrazol-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A386851-1G A386851-250MG A386851-100MG A386851-5G
76773-636EA 221 USD
76773-636 76773-638 77878-120 77878-121
5-Methyl-1-phenyl-1H-pyrazole-4-boronic acid, pinacol ester 98%
5-Methyl-1-phenyl-1H-pyrazole-4-boronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₆H₂₁BN₂O₂
MDL Number: MFCD06797463
CAS Number: 849776-88-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR