Bortezomib 98%

Supplier: Ambeed
Danger

Synonyms: ((R)-3-Methyl-1-((S)-3-phenyl-2-(pyrazine-2-carboxamido)propanamido)butyl)boronic acid

A385479-100MG A385479-1G A385479-1MG A385479-250MG A385479-5MG A385479-10MG A385479-50MG
76773-618EA 44.56 USD
76773-618 76773-620 76773-622 76773-624 76773-626 77551-136 77626-762
Bortezomib 98%
Bortezomib

Precaución: for Research Use Only

Formula: C₁₉H₂₅BN₄O₄
MW: 384.24 g/mol
MDL Number: MFCD09056737
CAS Number: 179324-69-7
UN: 3249
ADR: 6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR