(5-Amino-1,3-phenylene)dimethanol 95%

Supplier: Ambeed
Warning

A385184-100MG A385184-250MG A385184-1G
76773-614EA 11.67 USD
76773-614 76773-616 77551-130
(5-Amino-1,3-phenylene)dimethanol 95%
(5-Amino-1,3-phenylene)dimethanol

Precaución: for Research Use Only

Formula: C₈H₁₁NO₂
MW: 153.18 g/mol
MDL Number: MFCD08690090
CAS Number: 71176-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR