4-Acetoxy-2,5-dimethyl-3(2H)-furanone 98%

Supplier: Ambeed
Warning

Synonyms: Acetic Acid 4,5-Dihydro-2,5-dimethyl-4-oxofuran-3-yl Ester, 4,5-Dihydro-2,5-dimethyl-4-oxofuran-3-yl Acetate, Furaneol Acetate, 2,5-Dimethyl-4-oxo-4,5-dihydrofuran-3-yl acetate

A384509-10G A384509-1G A384509-25G A384509-5G
76773-606EA 11.67 USD
76773-606 76773-608 76773-610 76773-612
4-Acetoxy-2,5-dimethyl-3(2H)-furanone 98%
4-Acetoxy-2,5-dimethyl-3(2H)-furanone

Precaución: for Research Use Only

Formula: C₈H₁₀O₄
MW: 170.16 g/mol
MDL Number: MFCD00799462
CAS Number: 4166-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR