2,3,5-Trifluorobenzoic acid 98%
Supplier: Ambeed
|
Warning
|
A384252-1G
A384252-5G
A384252-250MG
76773-596EA
18
USD
76773-596
76773-598
77878-108
2,3,5-Trifluorobenzoic acid 98%
2,3,5-Trifluorobenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₃F₃O₂ |
MDL Number:
MFCD00061200 CAS Number: 654-87-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...