4-Chloro-2-methoxyaniline hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-o-anisidine hydrochloride

A383566-10G A383566-1G A383566-5G
76773-590EA 144.29 USD
76773-590 76773-592 76773-594
4-Chloro-2-methoxyaniline hydrochloride 97%
4-Chloro-2-methoxyaniline hydrochloride

Precaución: for Research Use Only

Formula: C₇H₉Cl₂NO
MW: 194.06 g/mol
MDL Number: MFCD06801386
CAS Number: 861299-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR