2-Fluoro-4-iodotoluene 98%

Supplier: Ambeed
Warning

A382153-100G A382153-10G A382153-25G A382153-5G
76773-580EA 90.18 USD
76773-580 76773-582 76773-584 76773-586
2-Fluoro-4-iodotoluene 98%
2-Fluoro-4-iodotoluene

Precaución: for Research Use Only

Formula: C₇H₆FI
MDL Number: MFCD00001045
CAS Number: 39998-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR