4-Benzoylpiperidine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: Phenyl(piperidin-4-yl)methanone hydrochloride

A380246-1G A380246-5G A380246-25G
76773-568EA 33 USD
76773-568 76773-570 77878-101
4-Benzoylpiperidine hydrochloride 98%
4-Benzoylpiperidine hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₆ClNO
MW: 225.7200 g/mol
MDL Number: MFCD00066982
CAS Number: 25519-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR