5-Bromo-2-methyl-4-nitro-1H-imidazole 95%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-methyl-4-nitroimidazole

A378243-1G A378243-250MG A378243-5G
76773-544EA 23.34 USD
76773-544 76773-546 76773-548
5-Bromo-2-methyl-4-nitro-1H-imidazole 95%
5-Bromo-2-methyl-4-nitro-1H-imidazole

Precaución: for Research Use Only

Formula: C₄H₄BrN₃O₂
MW: 205.997 g/mol
MDL Number: MFCD00156130
CAS Number: 18874-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR