5-Methyl-1,2,4-oxadiazol-3-amine 98%

Supplier: Ambeed
Warning

A376569-100MG A376569-1G A376569-250MG A376569-5G
76773-526EA 20 USD
76773-526 76773-528 76773-530 76773-532
5-Methyl-1,2,4-oxadiazol-3-amine 98%
5-Methyl-1,2,4-oxadiazol-3-amine

Precaución: for Research Use Only

Formula: C₃H₅N₃O
MDL Number: MFCD08272077
CAS Number: 40483-47-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR