(6-Bromopyridin-2-yl)methanamine 98%

Supplier: Ambeed
Danger

A375603-100MG A375603-1G A375603-250MG A375603-5G
76773-504EA 46.68 USD
76773-504 76773-506 76773-508 76773-510
(6-Bromopyridin-2-yl)methanamine 98%
(6-Bromopyridin-2-yl)methanamine

Precaución: for Research Use Only

Formula: C₆H₇BrN₂
MW: 187.03 g/mol
MDL Number: MFCD09261221
CAS Number: 188637-63-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR