Dimethyl 6-chloropyridine-2,3-dicarboxylate 96%

Supplier: Ambeed
Warning

A373824-100MG A373824-250MG A373824-1G
76773-488EA 51.99 USD
76773-488 76773-490 77601-652
Dimethyl 6-chloropyridine-2,3-dicarboxylate 96%
Dimethyl 6-chloropyridine-2,3-dicarboxylate

Precaución: for Research Use Only

Formula: C₉H₈ClNO₄
MW: 229.61 g/mol
MDL Number: MFCD04038569
CAS Number: 32383-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR