Bis(1H-benzo[d][1,2,3]triazol-5-yl)methane 97%
Supplier: Ambeed
|
Danger
|
A373474-10G
A373474-1G
A373474-250MG
A373474-5G
76773-476EA
112.46
USD
76773-476
76773-478
76773-480
76773-482
Bis(1H-benzo[d][1,2,3]triazol-5-yl)methane 97%
Bis(1H-benzo[d][1,2,3]triazol-5-yl)methane
Precaución: for Research Use Only
Formula:
C₁₃H₁₀N₆ MW: 250.25 g/mol |
MDL Number:
MFCD00758857 CAS Number: 15805-10-4 UN: 1325 ADR: 4.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...