6-Chloro-5-fluoro-2-oxindole 97%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-5-fluoroindolin-2-one

A373378-1G A373378-250MG A373378-5G
76773-472EA 58.35 USD
76773-472 76773-474 77594-114
6-Chloro-5-fluoro-2-oxindole 97%
6-Chloro-5-fluoro-2-oxindole

Precaución: for Research Use Only

Formula: C₈H₅ClFNO
MW: 185.583 g/mol
MDL Number: MFCD09839325
CAS Number: 100487-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR