5-Chloro-2-fluorobenzotrifluoride 95%

Supplier: Ambeed
Danger

Synonyms: 1-Chloro-4-fluoro-3-(trifluoromethyl)benzene, 2-Fluoro-5-chlorobenzotrifluoride, 4-Chloro-1-fluoro-2-(trifluoromethyl)benzene, 5-Chloro-2-fluorobenzotrifluoride

A371310-1G A371310-5G
76773-468EA 10.61 USD
76773-468 76773-470
5-Chloro-2-fluorobenzotrifluoride 95%
5-Chloro-2-fluorobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₃ClF₄
MW: 198.55 g/mol
MDL Number: MFCD00060655
CAS Number: 89634-74-2
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR