tert-Butyl-3-amino-4,6-dihydropyrrolo[3,4-c]pyrazole-5(1H)-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-1,4,5,6-tetrahydropyrrolo[3,4-c]pyrazole, N5-BOC protected, tert-Butyl 3-amino-4,6-dihydropyrrolo[3,4-c]pyrazole-5(1H)-carboxylate

A369336-1G A369336-250MG A369336-25G A369336-5G
76773-448EA 20.16 USD
76773-448 76773-450 76773-452 76773-454
tert-Butyl-3-amino-4,6-dihydropyrrolo[3,4-c]pyrazole-5(1H)-carboxylate 98%
tert-Butyl-3-amino-4,6-dihydropyrrolo[3,4-c]pyrazole-5(1H)-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₆N₄O₂
MW: 224.2596 g/mol
MDL Number: MFCD08275050
CAS Number: 398491-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR