7-Fluoroindole 98%

Supplier: Ambeed
Warning

A368722-100G A368722-10G A368722-1G A368722-250MG A368722-25G A368722-5G
76773-436EA 950.57 USD
76773-436 76773-438 76773-440 76773-442 76773-444 76773-446
7-Fluoroindole 98%
7-Fluoroindole

Precaución: for Research Use Only

Formula: C₈H₆FN
MW: 135.14 g/mol
MDL Number: MFCD01074502
CAS Number: 387-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR