2,6-Dibromo-p-toluidine 95%

Supplier: Ambeed
Warning

A368268-100G A368268-25G A368268-500G A368268-5G
76773-420EA 66 USD
76773-420 76773-422 76773-424 76773-426
2,6-Dibromo-p-toluidine 95%
2,6-Dibromo-p-toluidine

Precaución: for Research Use Only

Formula: C₇H₇Br₂N
MDL Number: MFCD00007641
CAS Number: 6968-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR