5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrin 98%

Supplier: Ambeed
Warning

Synonyms: 5,10,15,20-Tetrakis(4-methoxyphenyl)-21H,23H-porphine

A368203-1G A368203-25G A368203-5G
76773-414EA 32 USD
76773-414 76773-416 76773-418
5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrin 98%
5,10,15,20-Tetrakis(4-methoxyphenyl)porphyrin

Precaución: for Research Use Only

Formula: C₄₈H₃₈N₄O₄
MW: 734.86 g/mol
MDL Number: MFCD00012072
CAS Number: 22112-78-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR