4-Amino-2-hydroxypyrimidine-5-carboxylic acid 95%

Supplier: Ambeed
Warning

A366000-1G A366000-250MG
76773-396EA 333.13 USD
76773-396 76773-398
4-Amino-2-hydroxypyrimidine-5-carboxylic acid 95%
4-Amino-2-hydroxypyrimidine-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₅N₃O₃
MW: 155.11 g/mol
MDL Number: MFCD00047368
CAS Number: 3650-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR