6-Fluoro-3-formylchromone 98%

Supplier: Ambeed
Warning

Synonyms: 6-Fluorochromone-3-carboxaldehyde, 6-Fluoro-4-oxo-4H-chromene-3-carbaldehyde

A363098-10G A363098-1G A363098-250MG A363098-5G
76773-366EA 117.76 USD
76773-366 76773-368 76773-370 76773-372
6-Fluoro-3-formylchromone 98%
6-Fluoro-3-formylchromone

Precaución: for Research Use Only

Formula: C₁₀H₅FO₃
MW: 192.15 g/mol
MDL Number: MFCD00139060
CAS Number: 69155-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR