(2R,3S,4R,5S,6S)-6-Methyltetrahydro-2H-pyran-2,3,4,5-tetraol 95%

Supplier: Ambeed
Warning

A360717-1G A360717-5G
76773-344EA 33.95 USD
76773-344 76773-346
(2R,3S,4R,5S,6S)-6-Methyltetrahydro-2H-pyran-2,3,4,5-tetraol 95%
(2R,3S,4R,5S,6S)-6-Methyltetrahydro-2H-pyran-2,3,4,5-tetraol

Precaución: for Research Use Only

Formula: C₆H₁₂O₅
MW: 164.15 g/mol
MDL Number: MFCD00069812
CAS Number: 6696-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR