3-Bromo-5-(methoxycarbonyl)benzoic acid 95%

Supplier: Ambeed
Warning

A359197-100MG A359197-1G A359197-250MG A359197-5G
76773-336EA 9 USD
76773-336 76773-338 76773-340 76773-342
3-Bromo-5-(methoxycarbonyl)benzoic acid 95%
3-Bromo-5-(methoxycarbonyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₇BrO₄
MDL Number: MFCD06797598
CAS Number: 161796-10-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR