5-Bromo-2-isopropoxybenzonitrile 97%

Supplier: Ambeed
Danger

A354069-1G A354069-250MG A354069-5G
76773-302EA 12 USD
76773-302 76773-304 76773-306
5-Bromo-2-isopropoxybenzonitrile 97%
5-Bromo-2-isopropoxybenzonitrile

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrNO
MDL Number: MFCD02257441
CAS Number: 515832-52-7
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR