5-Chloro-2-(trifluoromethyl)phenol 98%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-4-(trifuloromethyl)phenol, 1-Chlor-2-hydroxy-5-(trifluormethyl)benzol, 3-Chloro-4-hydroxybenzotrifluoride, 2-Chloro-4-(trifluoromethyl)phenol

A352727-100G A352727-25G A352727-5G
76773-296EA 137.01 USD
76773-296 76773-298 76773-300
5-Chloro-2-(trifluoromethyl)phenol 98%
5-Chloro-2-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₇H₄ClF₃O
MDL Number: MFCD00042217
CAS Number: 35852-58-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR