2-Isobutyl-4-methylthiazole 98%

Supplier: Ambeed
Warning

A352588-25G A352588-5G
76773-292EA 25 USD
76773-292 76773-294
2-Isobutyl-4-methylthiazole 98%
2-Isobutyl-4-methylthiazole

Precaución: for Research Use Only

Formula: C₈H₁₃NS
MDL Number: MFCD00053138
CAS Number: 61323-24-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR