5-Bromo-2,2'-bithiophene-5'-carboxaldehyde 98%

Supplier: Ambeed
Danger

A352561-1G A352561-250MG A352561-5G
76773-286EA 14 USD
76773-286 76773-288 76773-290
5-Bromo-2,2'-bithiophene-5'-carboxaldehyde 98%
5-Bromo-2,2'-bithiophene-5'-carboxaldehyde

Precaución: for Research Use Only

Formula: C₉H₅BrOS₂
MDL Number: MFCD01321134
CAS Number: 110046-60-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR