Chlorpheniramine maleate 99%

Supplier: Ambeed
Warning

Synonyms: Chlorphenamine maleate

A352417-100G A352417-250MG A352417-25G A352417-500G A352417-5G A352417-100MG
76773-274EA 50 USD
76773-274 76773-276 76773-278 76773-280 76773-282 76773-284 77878-065
Chlorpheniramine maleate 99%
Chlorpheniramine maleate

Precaución: for Research Use Only

Formula: C₁₆H₁₉ClN₂
MW: 390.86 g/mol
MDL Number: MFCD00069225
CAS Number: 113-92-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR