2-Fluoro-6-methoxybenzonitrile 95%

Supplier: Ambeed
Warning

A349233-100G A349233-10G A349233-1G A349233-25G
76773-270EA 54 USD
76773-270 76773-272 77626-666 77626-668
2-Fluoro-6-methoxybenzonitrile 95%
2-Fluoro-6-methoxybenzonitrile

Precaución: for Research Use Only

Formula: C₈H₆FNO
MDL Number: MFCD00042291
CAS Number: 94088-46-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR