Manganese(II) chloride 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Manganese dichloride
A347892-1MG
A347892-25MG
A347892-5MG
A347892-100G
76773-256EA
5.31
USD
76773-256
76773-258
76773-260
77598-442
Manganese(II) chloride 98%
Manganese(II) chloride
Precaución: for Research Use Only
Formula:
MnCl₂ MW: 125.84 g/mol |
MDL Number:
MFCD00011114 CAS Number: 7773-01-5 UN: 3077 ADR: 9,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...