Isobutyl gallate 97%

Supplier: Ambeed
Warning

Synonyms: Gallic Acid Isobutyl Ester, Isobutyl 3,4,5-trihydroxybenzoate

A347096-1G A347096-25G A347096-5G
76773-246EA 24 USD
76773-246 76773-248 76773-250
Isobutyl gallate 97%
Isobutyl gallate

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₅
MW: 226.23 g/mol
MDL Number: MFCD00016425
CAS Number: 3856-05-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR