N-Methyl-7H-purin-6-amine 97%

Supplier: Ambeed
Warning

Synonyms: N6-Methyladenine, 6-Methylaminopurine

A345563-100MG A345563-1G A345563-250MG A345563-5G
76773-234EA 29.71 USD
76773-234 76773-236 76773-238 76773-240
N-Methyl-7H-purin-6-amine 97%
N-Methyl-7H-purin-6-amine

Precaución: for Research Use Only

Formula: C₆H₇N₅
MW: 149.16 g/mol
MDL Number: MFCD00075820
CAS Number: 443-72-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR